Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100664.5 | NP_172268.1 | 2 | 503 | 119 | |
GenBank mRNA | AB473841.1 | - | 2 | 451 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |