Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084776.2 | NP_001078245.1 | 3 | 750 | 109 | |
NM_114428.4 | NP_190145.1 | 2 | 827 | 109 | ||
GenBank mRNA | AK221492.1 | - | 3 | 697 | 109 | |
BT024625.1 | - | 1 | 564 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |