Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HWS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340111.1 | NP_001319816.1 | 2 | 1817 | 76 | |
NM_001340112.1 | NP_001327207.1 | 2 | 1817 | 76 | ||
NM_001340113.1 | NP_001327208.1 | 2 | 1887 | 76 | ||
NM_116024.3 | NP_191718.1 | 2 | 1670 | 76 | ||
GenBank mRNA | AF386994.1 | - | 2 | 1601 | 76 | |
BT000054.1 | - | 1 | 1223 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |