Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121157.2 | NP_196680.1 | 2 | 589 | 146 | |
GenBank mRNA | AY560865.1 | - | 2 | 437 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |