Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121569.3 | NP_197069.1 | 4 | 1264 | 158 | |
GenBank mRNA | AY039846.1 | - | 4 | 1248 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |