Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203423.1 | NP_001190352.1 | 2 | 1978 | 120 | |
NM_001343696.1 | NP_001331436.1 | 2 | 1841 | 120 | ||
NM_122112.4 | NP_568408.1 | 2 | 1856 | 120 | ||
GenBank mRNA | AY048254.1 | - | 2 | 1822 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |