Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101833.4 | NP_173407.1 | 2 | 1235 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |