Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125703.4 | NP_201114.1 | 2 | 1022 | 161 | |
NM_180921.2 | NP_851252.1 | 2 | 972 | 161 | ||
NM_180922.1 | NP_851253.1 | 3 | 1031 | 161 | ||
NM_180923.1 | NP_851254.1 | 3 | 1102 | 161 | ||
GenBank mRNA | AF447897.1 | - | 2 | 972 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |