Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126521.3 | NP_178566.1 | 2 | 1262 | 97 | |
GenBank mRNA | AF210732.1 | - | 2 | 1039 | 97 | |
AK175210.1 | - | 2 | 1012 | 97 | ||
BT026455.1 | - | 1 | 855 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |