Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUVH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336547.1 | NP_001324389.1 | 3 | 2767 | 119 | |
NM_001336548.1 | NP_001324388.1 | 2 | 3190 | 119 | ||
NM_001336549.1 | NP_001324386.1 | 2 | 3176 | 119 | ||
NM_001336550.1 | NP_001324387.1 | 3 | 2646 | 119 | ||
NM_129070.3 | NP_181061.1 | 3 | 2780 | 119 | ||
GenBank mRNA | AY062735.1 | - | 3 | 2730 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |