Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337215.1 | NP_001318436.1 | 4 | 1130 | 85 | |
NM_001337216.1 | NP_001324595.1 | 3 | 1516 | 85 | ||
NM_001337217.1 | NP_001324594.1 | 2 | 1352 | 85 | ||
NM_130246.3 | NP_182203.1 | 2 | 674 | 85 | ||
NM_180128.3 | NP_850459.1 | 3 | 966 | 85 | ||
GenBank mRNA | AK226483.1 | - | 2 | 554 | 85 | |
AK317164.1 | - | 3 | 603 | 85 | ||
AY086033.1 | - | 2 | 585 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |