Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180170.3 | NP_850501.1 | 2 | 1524 | 138 | |
GenBank mRNA | AB014465.1 | - | 2 | 1072 | 138 | |
BT001982.1 | - | 2 | 1120 | 138 | ||
BT008839.1 | - | 1 | 975 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |