Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125703.4 | NP_201114.1 | 2 | 881 | 59 | |
NM_180921.2 | NP_851252.1 | 2 | 831 | 59 | ||
NM_180922.1 | NP_851253.1 | 3 | 890 | 59 | ||
NM_180923.1 | NP_851254.1 | 3 | 961 | 59 | ||
GenBank mRNA | AB164305.1 | - | 1 | 517 | 59 | |
AB473837.1 | - | 2 | 806 | 59 | ||
AF447897.1 | - | 2 | 831 | 59 | ||
BT025745.1 | - | 1 | 517 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |