Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036070.1 | NP_001031147.1 | 1 | 1572 | 90 | |
NM_001333232.1 | NP_001319164.1 | 2 | 1934 | 90 | ||
NM_001333234.1 | NP_001322785.1 | 3 | 1816 | 90 | ||
NM_103556.4 | NP_175096.1 | 2 | 1377 | 90 | ||
NM_202243.3 | NP_973972.1 | 3 | 1644 | 90 | ||
GenBank mRNA | AY062782.1 | - | 2 | 1315 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |