Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF4-L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105922.4 | NP_565044.1 | 2 | 841 | 102 | |
GenBank mRNA | AY039916.1 | - | 2 | 752 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |