Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112968.4 | NP_188713.1 | 2 | 2265 | 122 | |
GenBank mRNA | AF004216.1 | - | 2 | 2155 | 122 | |
AY069875.2 | - | 2 | 2234 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |