Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAHH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113286.3 | NP_189023.1 | 2 | 1612 | 79 | |
GenBank mRNA | AY059888.1 | - | 2 | 1565 | 79 | |
AY093385.1 | - | 2 | 1529 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |