Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP71B20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113525.4 | NP_189249.1 | 2 | 1756 | 132 | |
NM_202635.1 | NP_974364.1 | 3 | 1327 | 132 | ||
GenBank mRNA | AY056270.1 | - | 2 | 1664 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |