Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT3G53730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115233.3 | NP_190941.1 | 1 | 912 | 92 |
Gene Symbol: HIS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY085175.1 | - | 1 | 429 | 91 | |
AY099675.1 | - | 1 | 431 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |