Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKE;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125756.5 | NP_201166.1 | 2 | 1767 | 93 | |
GenBank mRNA | AY600243.1 | - | 2 | 1610 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |