Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036140.1 | NP_001031217.1 | 1 | 1149 | 121 | |
NM_104833.3 | NP_176347.1 | 3 | 1153 | 121 | ||
GenBank mRNA | AY059879.1 | - | 3 | 982 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |