Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036555.3 | NP_001031632.1 | 7 - 8 | 537 | 114 | |
NM_001340903.1 | NP_001319932.1 | 8 - 9 | 755 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |