Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160880.2 | NP_001154352.1 | 2 - 3 | 1131 | 132 | |
NM_001198111.2 | NP_001185040.1 | 2 - 3 | 1068 | 132 | ||
NM_001332403.1 | NP_001319044.1 | 1 - 2 | 878 | 132 | ||
NM_001332404.1 | NP_001322544.1 | 2 - 3 | 1032 | 132 | ||
NM_101826.3 | NP_173400.1 | 2 - 3 | 1029 | 132 | ||
GenBank mRNA | AY570508.1 | - | 1 - 2 | 813 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |