Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036217.1 | NP_001031294.1 | 2 - 3 | 301 | 156 | |
NM_106393.5 | NP_565156.1 | 3 - 4 | 434 | 156 | ||
GenBank mRNA | AF043533.1 | - | 543 | 156 | ||
AF385730.1 | - | 3 - 4 | 351 | 156 | ||
AY081735.1 | - | 2 - 3 | 301 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |