Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124831.3 | NP_200262.1 | 1 - 2 | 445 | 116 | |
GenBank mRNA | AB050596.1 | - | 1 - 2 | 321 | 116 | |
BT021108.1 | - | 1 - 2 | 336 | 116 | ||
BT046117.1 | - | 1 - 2 | 321 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |