Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126443.5 | NP_178491.1 | 2 - 3 | 865 | 129 | |
GenBank mRNA | AK227153.1 | - | 2 - 3 | 797 | 129 | |
BT022022.1 | - | 2 - 3 | 804 | 129 | ||
BT026371.1 | - | 2 - 3 | 795 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |