Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036790.2 | NP_001031867.1 | 7 | 1340 | 139 | |
NM_001085095.1 | NP_001078564.1 | 8 | 1284 | 139 | ||
NM_001343104.1 | NP_001318526.1 | 8 | 1936 | 139 | ||
NM_001343105.1 | NP_001330918.1 | 7 | 2085 | 139 | ||
NM_001343106.1 | NP_001330917.1 | 7 | 2073 | 139 | ||
NM_001343107.1 | NP_001330916.1 | 6 | 1456 | 139 | ||
NM_001343108.1 | NP_001330915.1 | 7 | 1332 | 139 | ||
NM_121071.4 | NP_568226.1 | 8 | 2285 | 139 | ||
GenBank mRNA | AK317485.1 | - | 7 | 1316 | 139 | |
AY050832.1 | - | 8 | 1399 | 139 |
Gene Symbol: HISN6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124112.2 | NP_001117584.1 | 7 | 1311 | 139 | |
NM_001160995.1 | NP_001154467.1 | 8 | 1285 | 139 | ||
NM_001334509.1 | NP_001319364.1 | 8 | 1311 | 139 | ||
NM_001334510.1 | NP_001321654.1 | 6 | 1404 | 139 | ||
NM_001334511.1 | NP_001321653.1 | 6 | 1392 | 139 | ||
NM_105850.3 | NP_177337.1 | 8 | 1393 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |