Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126130.2 | NP_201531.1 | 1 | 946 | 74 | |
GenBank mRNA | AF326877.1 | - | 1 | 943 | 74 | |
AF339698.1 | - | 1 | 859 | 74 | ||
AY519648.1 | - | 1 | 859 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |