Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTFL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085029.3 | NP_001078498.1 | 1 | 252 | 95 | |
GenBank mRNA | DQ487702.1 | - | 1 | 46 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |