Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103668.3 | NP_564514.1 | 1 | 649 | 141 | |
GenBank mRNA | AK227048.1 | - | 1 | 650 | 141 | |
BT024474.1 | - | 1 | 624 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |