Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046465.1 | NP_001039930.1 | 6 - 7 | 811 | 71 | |
GenBank mRNA | BC113293.1 | - | 6 - 7 | 811 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |