Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076295.2 | NP_001069763.2 | 5 - 6 | 676 | 65 | |
GenBank mRNA | BC102911.1 | - | 5 - 6 | 677 | 65 | |
EH156727.1 | - | 5 - 6 | 672 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |