Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174282.3 | NP_776707.2 | 2 - 3 | 105 | 74 | |
GenBank mRNA | BC102358.1 | - | 2 - 3 | 115 | 74 | |
BT020662.1 | - | 2 - 3 | 105 | 74 | ||
BT020725.1 | - | 2 - 3 | 104 | 74 | ||
X75935.1 | - | 119 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |