Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104986.1 | NP_001098456.1 | 9 - 10 | 783 | 98 | |
GenBank mRNA | BC151395.1 | - | 9 - 10 | 783 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |