Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174815.2 | NP_777240.1 | 4 - 5 | 492 | 67 | |
GenBank mRNA | BC105363.1 | - | 4 - 5 | 492 | 67 | |
M95793.1 | - | 4 - 5 | 464 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |