Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191476.2 | NP_001178405.1 | 4 - 5 | 1221 | 96 | |
GenBank mRNA | EV635295.1 | - | 3 - 4 | 379 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |