Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100371.1 | NP_001093841.1 | 4 - 5 | 847 | 71 | |
GenBank mRNA | BC149232.1 | - | 4 - 5 | 847 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |