Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034530.2 | NP_001029702.1 | 2 - 3 | 378 | 125 | |
GenBank mRNA | BC102895.1 | - | 2 - 3 | 380 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |