Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075515.2 | NP_001068983.2 | 3 - 4 | 233 | 83 | |
GenBank mRNA | BC118394.1 | - | 3 - 4 | 209 | 83 | |
EE241538.1 | - | 3 - 4 | 234 | 83 | ||
EH136301.1 | - | 3 - 4 | 229 | 83 | ||
EH208684.1 | - | 3 - 4 | 198 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |