Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024523.1 | NP_001019694.1 | 1 - 2 | 327 | 135 | |
GenBank mRNA | BC102674.1 | - | 1 - 2 | 184 | 135 | |
BT020970.1 | - | 1 - 2 | 327 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |