Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011674.1 | NP_001011674.1 | 9 - 10 | 898 | 95 | |
GenBank mRNA | AB198072.1 | - | 9 - 10 | 898 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Bos taurus Probe spans exons |