Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002886.1 | NP_001002886.1 | 1 - 2 | 163 | 150 | |
GenBank mRNA | AY234181.1 | - | 1 - 2 | 163 | 150 | |
BC114794.1 | - | 1 - 2 | 133 | 150 | ||
DQ324373.1 | - | 1 - 2 | 159 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |