Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002887.3 | NP_001002887.3 | 2 - 3 | 220 | 76 | |
GenBank mRNA | AY427787.1 | - | 218 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |