Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003902.1 | NP_001003902.1 | 2 - 3 | 294 | 131 | |
GenBank mRNA | AY675077.1 | - | 2 - 3 | 294 | 131 | |
BC108164.1 | - | 2 - 3 | 220 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |