Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007813.2 | NP_001007814.1 | 5 - 6 | 725 | 89 | |
GenBank mRNA | AJ854251.1 | - | 5 - 6 | 727 | 89 | |
BC151789.1 | - | 5 - 6 | 739 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |