Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007818.1 | NP_001007819.1 | 2 - 3 | 274 | 74 | |
GenBank mRNA | AF529136.1 | - | 2 - 3 | 274 | 74 | |
BC103291.1 | - | 2 - 3 | 271 | 74 | ||
BT021545.1 | - | 2 - 3 | 270 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |