Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011677.1 | NP_001011677.1 | 4 - 5 | 763 | 121 | |
GenBank mRNA | AY841904.1 | - | 4 - 5 | 763 | 121 | |
BC102855.1 | - | 4 - 5 | 712 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |