Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011679.1 | NP_001011679.1 | 3 - 4 | 337 | 80 | |
GenBank mRNA | AY854632.1 | - | 3 - 4 | 337 | 80 | |
BC112599.1 | - | 3 - 4 | 349 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |