Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BREH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012287.1 | NP_001012287.1 | 12 - 13 | 1442 | 65 | |
GenBank mRNA | AY369075.1 | - | 12 - 13 | 1442 | 65 | |
BC102781.1 | - | 12 - 13 | 1493 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |