Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013594.1 | NP_001013612.1 | 2 - 3 | 281 | 104 | |
GenBank mRNA | EF208024.1 | - | 2 - 3 | 274 | 104 | |
M16894.1 | - | 2 - 3 | 281 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Bos taurus Probe spans exons |